Learn More
AVANTOR MASTERFLEX (VWR LLC) Red tubing, 1/16""ID x 1/8""OD, 50 ft/pack
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9615002
Encompass
Ideal for dedicated or multiple lines¿color-coding provides quick and easy line identification Characteristics: Superior heat resistance, low compression set, odorless, and nontoxic. Translucent. Certifications: Elastomer is FDA-compliant (21 CFR 177.2600). Tubing is manufactured according to GMP. Temperature range: ¿60 to 460¿F (¿51 to 238¿C) Sterilize: Autoclave or gamma irradiation.
Catalog No. NC1124221