Learn More
MSC RED VINYL TAPE
Catalog No. 5000673701
Encompass
This masking tape is 36 yd. long and 5.2 mil thick, featuring a red color. It is made with vinyl material and a rubber adhesive, providing a tensile strength of 14 lb/"
  • Suitable for temporary surface protection
  • Ideal for wrapping or sealing applications
  • Supports die-cutting applications
  • Used for bundling and reinforcing
  • Effective for paint masking
  • Provides identification
  • Appropriate for anodizing and electroplating
  • Can be used for printable labels
  • Useful for floor lane marking
  • Excellent for color coding and decorating
  • Operating temperature range: 4°C to 77°C (40°F to 170°F)
Catalog No. 50-006-73701 Supplier MSC Supplier No. 60549383
This masking tape is 36 yd. long and 5.2 mil thick, featuring a red color. It is made with vinyl material and a rubber adhesive, providing a tensile strength of 14 lb/"
  • Suitable for temporary surface protection
  • Ideal for wrapping or sealing applications
  • Supports die-cutting applications
  • Used for bundling and reinforcing
  • Effective for paint masking
  • Provides identification
  • Appropriate for anodizing and electroplating
  • Can be used for printable labels
  • Useful for floor lane marking
  • Excellent for color coding and decorating
  • Operating temperature range: 4°C to 77°C (40°F to 170°F)