Learn More
Electron Microscopy Sciences Red-Z Fluid Control Solidifier 15 OZ
SDP
Catalog No. 5028325
Encompass_Preferred
RED-Z will solidify any water base fluid at a ratio of 3/4 oz to 1 full liter. Solidifies, decontaminates and deodorizers. Contains chlorine (NaDCC)an EPA registered disinfectant-almost twice the amount recommended by the CDC. An Alternate treatment to steam sterilization or incineration. For blood and other body fluids. Available in a pointed top bottle that fits easily in porthole containers and in a flip-top shaker top bottle for large spills.
Catalog No. 50-283-25 Supplier Electron Microscopy Sciences Supplier No. 2414525
RED-Z will solidify any water base fluid at a ratio of 3/4 oz to 1 full liter. Solidifies, decontaminates and deodorizers. Contains chlorine (NaDCC)an EPA registered disinfectant-almost twice the amount recommended by the CDC. An Alternate treatment to steam sterilization or incineration. For blood and other body fluids. Available in a pointed top bottle that fits easily in porthole containers and in a flip-top shaker top bottle for large spills.