Learn More
Cytek Biosciences Inc rF 710 aHu CD11c Anti Primary
Supplier: Cytek Biosciences Inc 800116T025
Product Name: redFluor 710 Anti-Human CD11c (3.9)
Product Class: Antibody
Type: Primary
Product Name: redFluor 710 Anti-Human CD11c (3.9)
Cytek Catalog Number: 80-0116-T025
Marker: CD11c
Target Species: Human
Clonality: Monoclonal
Clone: 3.9
Host Species: Mouse
Isotype: IgG1, k
Conjugation: redFluor 710
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.