Learn More
Biotium RedDot™2, 200X in DMSO
Catalog No. 501964654
Encompass_Preferred
RedDot™2 is a far-red cell membrane-impermeant nuclear dye similar to Draq7™. Ideal for staining the nuclei of dead or fixed and permeabilized cells with minimal cytoplasmic RNA staining. The dye is highly thermostable and photostable, providing convenient handling and ideal for demanding applications such as confocal microscopy. The dye has an excitation/emission of 665/695 nm when bound to DNA. Properties: Liquid; Store at 4°C, protect from light; Ex/Em= 665/695 nm (with DNA).
Catalog No. 50-196-4654 Supplier Biotium Supplier No. 400611
RedDot™2 is a far-red cell membrane-impermeant nuclear dye similar to Draq7™. Ideal for staining the nuclei of dead or fixed and permeabilized cells with minimal cytoplasmic RNA staining. The dye is highly thermostable and photostable, providing convenient handling and ideal for demanding applications such as confocal microscopy. The dye has an excitation/emission of 665/695 nm when bound to DNA. Properties: Liquid; Store at 4°C, protect from light; Ex/Em= 665/695 nm (with DNA).