Learn More
Biotium REDDOT2 FAR-RED NUCLEAR STAIN
Catalog No. NC0772106
Encompass_Preferred
RedDot™2 is a far-red cell membrane-impermeable nuclear dye similar to Draq7™ with excitation/emission at 665/695 nm. The dye is ideal for specifically staining the nuclei of dead cells with minimal cytoplasmic RNA staining. RedDot™2 provides excellent nuclear counterstaining in fixed and permeabilized cells and tissue sections without the need for RNase treatment to remove cytoplasmic RNA. The dye is highly thermostable and photostable, providing convenient handling and ideal for demanding applications such as confocal microscopy. RedDot™2 also has been used for cell number normalization for near-infrared In Cell Western™. The excitation maximum of RedDot™2 is 665 nm, but it can be efficiently excited by wavelengths from 488 to 647 nm, and therefore can be used with the 488 nm flow cytometry laser line. Supplied as a 200X stock solution in DMSO, 25 uL trial size. In Cell Western is a trademark of LI-COR® Biosciences.
Catalog No. NC0772106 Supplier Biotium Supplier No. 40061T
RedDot™2 is a far-red cell membrane-impermeable nuclear dye similar to Draq7™ with excitation/emission at 665/695 nm. The dye is ideal for specifically staining the nuclei of dead cells with minimal cytoplasmic RNA staining. RedDot™2 provides excellent nuclear counterstaining in fixed and permeabilized cells and tissue sections without the need for RNase treatment to remove cytoplasmic RNA. The dye is highly thermostable and photostable, providing convenient handling and ideal for demanding applications such as confocal microscopy. RedDot™2 also has been used for cell number normalization for near-infrared In Cell Western™. The excitation maximum of RedDot™2 is 665 nm, but it can be efficiently excited by wavelengths from 488 to 647 nm, and therefore can be used with the 488 nm flow cytometry laser line. Supplied as a 200X stock solution in DMSO, 25 uL trial size. In Cell Western is a trademark of LI-COR® Biosciences.