Learn More
Cayman Chemical RedDotM2 VIabIlIty Dye 50uL
SDP
Catalog No. 502893872
Encompass_Preferred
A p-phenylenediamine and non-oxidized version of DPPD-Q toxic to the aquatic bacterium V. fischeri (EC50 0.02 mg/L) reduces the viability of HepG2 cancer cells at 10 UG/L decreases urinary protein levels and serum levels of bilirubin in male rats at 100 mg/kg per day increases the time in gestation for female rats when administered to pregnant dams at 50 and 300 mg/kg per day has been found in water treatment plant influent biosolids and landfill leachates indoor dust and in PM2.5 samples collected in cities and by roadsides
Catalog No. 50-289-3872 Supplier Cayman Chemical Supplier No. 60128250 UL
A p-phenylenediamine and non-oxidized version of DPPD-Q toxic to the aquatic bacterium V. fischeri (EC50 0.02 mg/L) reduces the viability of HepG2 cancer cells at 10 UG/L decreases urinary protein levels and serum levels of bilirubin in male rats at 100 mg/kg per day increases the time in gestation for female rats when administered to pregnant dams at 50 and 300 mg/kg per day has been found in water treatment plant influent biosolids and landfill leachates indoor dust and in PM2.5 samples collected in cities and by roadsides