Learn More
Sigma Aldrich Fine Chemicals Biosciences REDExtract N AmpTM Plant P
Supplier:  Sigma Aldrich Fine Chemicals Biosciences XNAPR1KT
Encompass
The REDExtract-N-Amp Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. The REDExtract-N-Amp PCR ReadyMix contains a dye that acts as a tracking dye and allows for convenient direct loading of PCR reactions onto agarose gels for analysis.
Catalog No. 50-290-7845