Learn More
Sigma Aldrich Fine Chemicals Biosciences REDExtract N AmpTM Plant P
Catalog No. 502905437
Encompass
The REDExtract-N-Amp Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. The REDExtract-N-Amp PCR ReadyMix contains a dye that acts as a tracking dye and allows for convenient direct loading of PCR reactions onto agarose gels for analysis.
Catalog No. 50-290-5437 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. XNAP1KT
The REDExtract-N-Amp Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. The REDExtract-N-Amp PCR ReadyMix contains a dye that acts as a tracking dye and allows for convenient direct loading of PCR reactions onto agarose gels for analysis.