Learn More
Sigma Aldrich Fine Chemicals Biosciences REDExtt N AmpTM PCR Rea125ML
Catalog No. 502914726
Encompass
The REDExtract-N-Amp(TM) PCR reaction mix is a PCR Master mix containing buffer salts dNTPs and REDTaq DNA polymerase. This PCR ReadyMix is intended for use with Sigma Extract-N-Amp Plant PCR kit and Extract-N-Amp Tissue PCR Kit. All Extract-N-Amp kits are sufficient for one PCR amplification per extraction. However where additional PCR reactions are required this supplemental PCR ReadyMix may be needed. The REDExtract-N-Amp PCR ReadyMix contains an inert tracking dye that allows for convenient direct loading of PCR products onto agarose gels for analysis.
Catalog No. 50-291-4726 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R4775125ML
Add to Cart
Add to Cart
Request a Quote
The REDExtract-N-Amp(TM) PCR reaction mix is a PCR Master mix containing buffer salts dNTPs and REDTaq DNA polymerase. This PCR ReadyMix is intended for use with Sigma Extract-N-Amp Plant PCR kit and Extract-N-Amp Tissue PCR Kit. All Extract-N-Amp kits are sufficient for one PCR amplification per extraction. However where additional PCR reactions are required this supplemental PCR ReadyMix may be needed. The REDExtract-N-Amp PCR ReadyMix contains an inert tracking dye that allows for convenient direct loading of PCR products onto agarose gels for analysis.