Learn More
Cytek Biosciences Inc Antibodies redFluor™ 710 Anti-Mouse CD48 (HM48-1) ….. Cat. No. 80-0481-U025 ….. Size 25 µg
Supplier:  Cytek Biosciences Inc 800481U025
Encompass_Preferred
Antibodies redFluor™ 710 Anti-Mouse CD48 (HM48-1) ….. Cat. No. 80-0481-U025 ….. Size 25 µgThe HM48-1 monoclonal antibody reacts with mouse CD48, a 45 kDa GPI-anchored glycoprotein that is also known as BCM1. CD48 is a member of the signaling lymphocyte activation molecule (SLAM) family and Ig superfamily. It is expressed on leukocytes, but not on non-hematopoietic cells and plays a role in cell adhesion and activation. The primary ligands for CD48 are CD2 (LFA-2) and CD244 (2B4 antigen). The HM48-1 antibody is reported to block in vitro and in vivo CD48 mediated interactions.
Catalog No. 50-210-3655