Learn More
Med Vet International Redi-Warm Fluid Bag Warmer, J0900R4T, Each
Catalog No. 5000584072
Encompass_Preferred
Additionally, emergency abdominal flushes can be administered in an instant with warm fluids on standby. The new Ready Warm fluid bag warmer maintains up to 4 1-liter IV bags at ready-to-use temperatures, with digital controls programmed to show the correct temperature of the fluids at all times. The bags can be left on the warmer for extended periods of time to be ready for emergencies. CONSTRUCTION Corrosion resistant aluminum Durable metal construction SPECIFICATIONS Temperature range: 122°F +/-5°F Preset at 100°F (37°C) Accuracy: +/- 1.8°F WEIGHT AND DIMS Length: 22.835" (580mm) Width: 12" (305mm) Height: 3.07" (78mm) Weight: 13lbs (5.89kg) POWER / CONTROLS Lighted power switch Auto-switching CE / cUL approved power adapter Output: 12vDC / 10A Input: 100-240vAC 50/60Hz Actual Weight: 14 lb. Weight reflected in additional information is in reference to dimensional weight for shipping purposes.
Catalog No. 50-005-84072 Supplier Med Vet International Supplier No. JOR900R4T
Additionally, emergency abdominal flushes can be administered in an instant with warm fluids on standby. The new Ready Warm fluid bag warmer maintains up to 4 1-liter IV bags at ready-to-use temperatures, with digital controls programmed to show the correct temperature of the fluids at all times. The bags can be left on the warmer for extended periods of time to be ready for emergencies. CONSTRUCTION Corrosion resistant aluminum Durable metal construction SPECIFICATIONS Temperature range: 122°F +/-5°F Preset at 100°F (37°C) Accuracy: +/- 1.8°F WEIGHT AND DIMS Length: 22.835" (580mm) Width: 12" (305mm) Height: 3.07" (78mm) Weight: 13lbs (5.89kg) POWER / CONTROLS Lighted power switch Auto-switching CE / cUL approved power adapter Output: 12vDC / 10A Input: 100-240vAC 50/60Hz Actual Weight: 14 lb. Weight reflected in additional information is in reference to dimensional weight for shipping purposes.