Learn More
MSC Boss Glove, Top Grain Goatskin, Kevlar Blend Lining, Hi-Vis Impact Protection, 1 Pair
Catalog No. 5000685325
Encompass
These drivers gloves are crafted from top grain goatskin leather to provide a balance of softness, abrasion resistance, and tactile sensitivity. They feature a Kevlar® blend fiber lining on the palm for cut protection and include thermo plastic rubber impact protection on the back of the hand and fingers to disperse impact force energy.
  • Top grain goatskin leather construction for durability and dexterity
  • Palm lining made with cut-resistant Kevlar® blend fiber
  • Back-of-hand and finger protection using thermo plastic rubber to mitigate impact
  • Secure hook and loop closure for easy on and off
  • Meets chromium VI free leather requirements
  • ANSI puncture level 4 and ANSI impact level 1
  • EN 388 performance results: 2X43FP
  • ANSI cut level A7
Catalog No. 50-006-85325 Supplier MSC Supplier No. 31129521
These drivers gloves are crafted from top grain goatskin leather to provide a balance of softness, abrasion resistance, and tactile sensitivity. They feature a Kevlar® blend fiber lining on the palm for cut protection and include thermo plastic rubber impact protection on the back of the hand and fingers to disperse impact force energy.
  • Top grain goatskin leather construction for durability and dexterity
  • Palm lining made with cut-resistant Kevlar® blend fiber
  • Back-of-hand and finger protection using thermo plastic rubber to mitigate impact
  • Secure hook and loop closure for easy on and off
  • Meets chromium VI free leather requirements
  • ANSI puncture level 4 and ANSI impact level 1
  • EN 388 performance results: 2X43FP
  • ANSI cut level A7