Learn More
Biolog Inc REDOX DYE MA
SDP
Catalog No. NC9071493
Encompass
Biolog Phenotype MicroArray Redox Dye MA for mammalian cells (6X concentration), 20ml for approximately 20 microplates
Catalog No. NC9071493 Supplier Biolog Inc Supplier No. 74351
Biolog Phenotype MicroArray Redox Dye MA for mammalian cells (6X concentration), 20ml for approximately 20 microplates