Learn More
Biolog Inc REDOX DYE MB
SDP
Catalog No. NC9071494
Encompass
Biolog Phenotype MicroArray Redox Dye MB for mammalian cells (6X concentration), 20ml for approximately 20 microplates
Catalog No. NC9071494 Supplier Biolog Inc Supplier No. 74352
Biolog Phenotype MicroArray Redox Dye MB for mammalian cells (6X concentration), 20ml for approximately 20 microplates