Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Redox standard 200 mV, MilliporeSigma™ Supelco™

Catalog No. 111017267 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7267 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7267 Supplier MilliporeSigma Supelco Supplier No. 40661500ML
Add to Cart
Add to Cart

The value is the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride

Cleaning Products, Cosmetics, Environmental, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 14459-95-1
Type Analytical Standard
Quantity 500 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.