Learn More
Sigma Aldrich Fine Chemicals Biosciences REDTaqR ReadyMixTM PCR Rea
Catalog No. 502900158
Encompass
REDTaq(R) ReadyMix(TM) is a ready-to-use mixture of Taq DNA polymerase 99% pure DNTPs reaction buffer and an inert red dye in a 2X concentrate. After the PCR reaction the PCR product can be loaded directly onto an agarose gel. The red loading dye migrates at approximately the same rate as a 125 base pair fragment in a 1% agarose gel. The inert dye has no effect on the amplification process and therefore a sample can be easily re-amplified such as in nested PCR.
Catalog No. 50-290-0158 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R252320RXN
REDTaq(R) ReadyMix(TM) is a ready-to-use mixture of Taq DNA polymerase 99% pure DNTPs reaction buffer and an inert red dye in a 2X concentrate. After the PCR reaction the PCR product can be loaded directly onto an agarose gel. The red loading dye migrates at approximately the same rate as a 125 base pair fragment in a 1% agarose gel. The inert dye has no effect on the amplification process and therefore a sample can be easily re-amplified such as in nested PCR.