Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Compression Reducer, 1/4"" OD x 1/8"" OD; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0638105
Encompass
Very good chemical resistance. Rates to 220 psi at 77¿F ( 25¿C). Usable from 32 to 212¿F (0 to 100¿C).
Catalog No. NC0850745