Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Tee, Hose Barb to Threaded Adapter, 1/4"" ID x 1/4"" ID x 3/8"" NPT(M); 10/PK
Catalog No. 50630601
Encompass
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
Catalog No. 50-630-601 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062412
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.