Learn More
Costech Analytical Technologies Inc Copper Wires, reduced 0.7 mm
Catalog No. NC9478521
Encompass_Preferred
Copper wires, 200 gm, Reduced 0.7 mm, Catalysts and Reagent for: Nitrogen/Protein Analysis;N, N/C and CHN
Catalog No. NC9478521 Supplier Costech Analytical Technologies Inc Supplier No. 011013
Copper wires, 200 gm, Reduced 0.7 mm, Catalysts and Reagent for: Nitrogen/Protein Analysis;N, N/C and CHN