Learn More
Acs Material Reduced Graphene Oxide (rGO) 1g
SDP
Catalog No. NC3793831
Encompass
The two-dimensional carbon allotrope known as graphene has become one of the most popular materials on the planet, and for good reason. With the ability to conduct massive amounts of heat and electricity along with its flexible and lightweight nature, graphene has already begun to improve the effectiveness of existing materials. Reduced graphene oxide can be achieved through a variety of chemical and electrochemical means. Scientists have been working on effective ways to reduce graphene oxide with the hope of being able to create large quantities of high-quality graphene that can be used in large-scale operations. While the process of making reduced graphene oxide hasn’t yet been perfected, this emerging material will surely shape the way industry leaders do business in the years to come.
Catalog No. NC3793831 Supplier Acs Material Supplier No. GNCR0001
The two-dimensional carbon allotrope known as graphene has become one of the most popular materials on the planet, and for good reason. With the ability to conduct massive amounts of heat and electricity along with its flexible and lightweight nature, graphene has already begun to improve the effectiveness of existing materials. Reduced graphene oxide can be achieved through a variety of chemical and electrochemical means. Scientists have been working on effective ways to reduce graphene oxide with the hope of being able to create large quantities of high-quality graphene that can be used in large-scale operations. While the process of making reduced graphene oxide hasn’t yet been perfected, this emerging material will surely shape the way industry leaders do business in the years to come.