Learn More
AVANTOR MASTERFLEX (VWR LLC) REDUCER 1/2IN X3/4IN PP 5/PK
Catalog No. 502742973
Encompass
Masterflex Fitting, Polypropylene, Straight, Hose Barb Reducer, 1/2IN ID x 3/4IN ID; 5/PK, Avantor
Catalog No. 50-274-2973 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636896
Masterflex Fitting, Polypropylene, Straight, Hose Barb Reducer, 1/2IN ID x 3/4IN ID; 5/PK, Avantor