Learn More
Grainger John Guest Speedfit Stem Reducer, 1 in. x 3/4 in., Engineered Plastic, 1 ea
Catalog No. 502768618
Encompass_Preferred
This stem reducer provides a dependable way to reduce the diameter of Speedfit fittings and connect Copper, PEX, or CPVC pipe in any combination. Designed with a twist-to-lock feature and an EPDM O-ring, it ensures a secure, leak-proof seal without the need for solder, glue, or tools. It is approved for behind-the-wall installations and suitable for hot and cold potable water and hydronic heating applications.
  • Connects copper, PEX, and CPVC pipe in any combination
  • Requires no tools, soldering, glue, or clamps
  • Approved for behind-the-wall installations without access panels
  • Features EPDM O-ring for a permanent, leak-proof seal
  • Rated for 160 psi at 73°F, 100 psi at 180°F, and 80 psi at 200°F
  • Lead-free construction
  • Backed by a 25-year warranty
Catalog No. 50-276-8618 Supplier Grainger Supplier No. 16T741
This stem reducer provides a dependable way to reduce the diameter of Speedfit fittings and connect Copper, PEX, or CPVC pipe in any combination. Designed with a twist-to-lock feature and an EPDM O-ring, it ensures a secure, leak-proof seal without the need for solder, glue, or tools. It is approved for behind-the-wall installations and suitable for hot and cold potable water and hydronic heating applications.
  • Connects copper, PEX, and CPVC pipe in any combination
  • Requires no tools, soldering, glue, or clamps
  • Approved for behind-the-wall installations without access panels
  • Features EPDM O-ring for a permanent, leak-proof seal
  • Rated for 160 psi at 73°F, 100 psi at 180°F, and 80 psi at 200°F
  • Lead-free construction
  • Backed by a 25-year warranty