Learn More
MSC Karcher Reducer, DN 40 to DN 35, Black, 1 ea
Catalog No. 502817644
Encompass
This reducer is designed for extending accessories for industrial NT vacuum cleaners. It allows for the combination of accessories with different connecting cone diameters, ensuring broader compatibility for various cleaning tasks.
  • Connects accessories with DN 35 and DN 40 connecting cones
  • Suitable for industrial NT vacuum cleaner range
  • Made of durable black plastic
  • Lightweight design for easy handling
  • Dimensions: 100 mm (L) × 44 mm (W) × 44 mm (H)
Catalog No. 50-281-7644 Supplier MSC Supplier No. 16617185
This reducer is designed for extending accessories for industrial NT vacuum cleaners. It allows for the combination of accessories with different connecting cone diameters, ensuring broader compatibility for various cleaning tasks.
  • Connects accessories with DN 35 and DN 40 connecting cones
  • Suitable for industrial NT vacuum cleaner range
  • Made of durable black plastic
  • Lightweight design for easy handling
  • Dimensions: 100 mm (L) × 44 mm (W) × 44 mm (H)