Learn More
MSC Karcher Reducer Adapter, DN 40 to DN 35, 1 ea
Catalog No. 502819047
Encompass
This Kärcher reducer adapter connects accessories with a DN 35 connection to accessory parts with a DN 40 connection. It is designed for use with industrial NT vacuum cleaners, facilitating compatibility between various components like upholstery nozzles, crevice nozzles, bends, and suction tubes.
  • Enables connection of DN 35 accessories to DN 40 parts
  • Ideal for industrial NT vacuum cleaner range
  • Standard nominal width 40 mm
  • Standard nominal width 2 35 mm
  • Color: black
  • Dimensions: 100 x 44 x 44 mm
Catalog No. 50-281-9047 Supplier MSC Supplier No. 16617136
This Kärcher reducer adapter connects accessories with a DN 35 connection to accessory parts with a DN 40 connection. It is designed for use with industrial NT vacuum cleaners, facilitating compatibility between various components like upholstery nozzles, crevice nozzles, bends, and suction tubes.
  • Enables connection of DN 35 accessories to DN 40 parts
  • Ideal for industrial NT vacuum cleaner range
  • Standard nominal width 40 mm
  • Standard nominal width 2 35 mm
  • Color: black
  • Dimensions: 100 x 44 x 44 mm