Learn More
Grainger John Guest Stem Reducer, Polypropylene, 3/4 in. x 1/2 in.
Catalog No. 502765256
Encompass_Preferred
The Speedfit Stem Reducer is designed to efficiently reduce the diameter of another Speedfit fitting. Constructed from durable polypropylene, this connector features Twist and Lock technology to provide a secure, leak-proof connection without the need for tools, soldering, glue, or clamps.
  • Features EPDM O-ring for reliable sealing
  • Compatible for use with copper, PEX, and CPVC piping in any combination
  • Approved for behind-the-wall applications without an access panel
  • Suitable for installation on wet lines
  • Rated for 160 psi at 73°F in water applications
  • Includes 25-year warranty
  • Lead free construction
Catalog No. 50-276-5256 Supplier Grainger Supplier No. 16T740
The Speedfit Stem Reducer is designed to efficiently reduce the diameter of another Speedfit fitting. Constructed from durable polypropylene, this connector features Twist and Lock technology to provide a secure, leak-proof connection without the need for tools, soldering, glue, or clamps.
  • Features EPDM O-ring for reliable sealing
  • Compatible for use with copper, PEX, and CPVC piping in any combination
  • Approved for behind-the-wall applications without an access panel
  • Suitable for installation on wet lines
  • Rated for 160 psi at 73°F in water applications
  • Includes 25-year warranty
  • Lead free construction