Learn More
Ideal Vacuum Products LLC REDUCER CONICAL KF-25 TO KF-40
SDP
Catalog No. NC1625545
Encompass
Reducer Conical KF-25 to KF-40 Vacuum Fittings, ISO-KF Flange Size NW-25 to NW-40, Stainless Steel
Catalog No. NC1625545 Supplier Ideal Vacuum Products LLC Supplier No. P101230
Reducer Conical KF-25 to KF-40 Vacuum Fittings, ISO-KF Flange Size NW-25 to NW-40, Stainless Steel