Learn More
Bellco Glass, Inc. Reducer Fitting Pack, Polyethylene, 1/2" HB x 1/4" HB, 10/PK
SDP
Catalog No. 502550499
Encompass_Preferred
Connectors are available in a variety of standard sizes. They provide a secure, watertight connection. These connectors make it possible to connect two hoses, securely and efficiently. Made from polyethylene. Material: PolyethyleneColor: BlackOD, 1/2" side: 0.545"OD, 1/4" side: 0.310"Length: 1.72"
Catalog No. 50-255-0499 Supplier Bellco Glass, Inc. Supplier No. 452800263
Connectors are available in a variety of standard sizes. They provide a secure, watertight connection. These connectors make it possible to connect two hoses, securely and efficiently. Made from polyethylene. Material: PolyethyleneColor: BlackOD, 1/2" side: 0.545"OD, 1/4" side: 0.310"Length: 1.72"