Learn More
Bellco Glass, Inc. Reducer Fitting Pack, Polyethylene, 5/8" Hose Barb x 1/2" Hose Barb, 100/PK
SDP
Supplier:  Bellco Glass, Inc. 452800271
Encompass_Preferred
Connectors are available in a variety of standard sizes. They provide a secure, watertight connection. These connectors make it possible to connect two hoses, securely and efficiently. Made from polyethylene. Material: PolyethyleneColor: BlackOD, 1/2" side: 0.545"OD, 5/8" side: 0.650"Length: 1.70"
Catalog No. 50-255-0491