Learn More
Bellco Glass, Inc. Reducer Fitting Pack, Polyethylene, 5/8" Hose Barb x 3/8" Hose Barb, 100/PK
SDP
Catalog No. 502550492
Encompass_Preferred
Connectors are available in a variety of standard sizes. They provide a secure, watertight connection. These connectors make it possible to connect two hoses, securely and efficiently. Made from polyethylene. Material: PolyethyleneColor: BlackOD, 3/8" side: 0.430"OD, 5/8" side: 0.650"Length: 1.70"
Catalog No. 50-255-0492 Supplier Bellco Glass, Inc. Supplier No. 452800270
Connectors are available in a variety of standard sizes. They provide a secure, watertight connection. These connectors make it possible to connect two hoses, securely and efficiently. Made from polyethylene. Material: PolyethyleneColor: BlackOD, 3/8" side: 0.430"OD, 5/8" side: 0.650"Length: 1.70"