Learn More
Bellco Glass, Inc. Reducer Fitting Pack, Polyethylene, 3/4" HB x 1/2" HB, 10/PK
SDP
Supplier:  Bellco Glass, Inc. 452800267
Encompass_Preferred
Connectors are available in a variety of standard sizes. They provide a secure, watertight connection. These connectors make it possible to connect two hoses, securely and efficiently. Made from polyethylene. Material: PolyethyleneColor: BlackOD, 1/2" side: 0.545"OD, 3/4" side: 0.800"Length: 2.91"
Catalog No. 50-255-0495