Learn More
Chemglass Life Sciences Adapter, Connecting, 24/40 Top Outer, 29/26 Lower Inner, Reducing
SDP
Catalog No. CG100046
Encompass_Preferred
Adapter, Connecting, 24/40 Top Outer, 29/26 Lower Inner, Reducing. Used to connect dissimilar standard taper joint sizes. Lower inner joint is larger in size than the top outer joint.
Catalog No. CG100046 Supplier Chemglass Life Sciences Supplier No. CG100046
Adapter, Connecting, 24/40 Top Outer, 29/26 Lower Inner, Reducing. Used to connect dissimilar standard taper joint sizes. Lower inner joint is larger in size than the top outer joint.