Learn More
Phenomenex Inc Reducing Adapter Assembly, PEEK
Catalog No. 502397221 Shop All Phenomenex Products
Encompass_Preferred
Reducing Adapter Assembly, PEEK joins 1/16" OD to 360µm OD tubing w/Fittings (MAX 5,000 psi / 345 bar)
Catalog No. 50-239-7221 Supplier Phenomenex Inc Supplier No. AN01023
Reducing Adapter Assembly, PEEK joins 1/16" OD to 360µm OD tubing w/Fittings (MAX 5,000 psi / 345 bar)