Learn More
Phenomenex Inc Reducing Adapter Assembly, SS
Catalog No. 502400626 Shop All Phenomenex Products
Encompass_Preferred
Reducing Adapter Assembly, SS joins 1/16" OD to 1/32 OD tubing w/Fittings (MAX 15,000 psi/ 1034 bar)
Catalog No. 50-240-0626 Supplier Phenomenex Inc Supplier No. AN01021
Reducing Adapter Assembly, SS joins 1/16" OD to 1/32 OD tubing w/Fittings (MAX 15,000 psi/ 1034 bar)