Learn More
Fritsch Milling And Sizing Inc Reducing Adapter for Grinding Bowls 12 - 80 Ml
SDP
Catalog No. 501534718
Encompass
Reducing adapter for grinding bowls 12 - 80 ml is necessary, if only one grinding bowl of 12, 25, 45 or 80 ml volume per grinding bowl holder is inserted in the PULVERISETTE 4, PULVERISETTE 5 or PULVERISETTE 6 classic line, these adapter is essential as adjustment.
Catalog No. 50-153-4718 Supplier Fritsch Milling And Sizing Inc Supplier No. 90.1120.09
Reducing adapter for grinding bowls 12 - 80 ml is necessary, if only one grinding bowl of 12, 25, 45 or 80 ml volume per grinding bowl holder is inserted in the PULVERISETTE 4, PULVERISETTE 5 or PULVERISETTE 6 classic line, these adapter is essential as adjustment.