Learn More
Grainger Reducing Bushing 316 SS 3/4 x 1/4 in
Catalog No. 502752204
Encompass_Preferred
The Parker Reducing Bushing is made from 316 stainless steel and has NPT thread connections. It reduces pipe size from 1" to 3/4". The bushing has a pressure rating of 3000 psi at 100°F. It can be used with Schedule 80 pipe and is manufactured in the USA. The bushing measures 1.38" in length and weighs 0.19 lbs.
Catalog No. 50-275-2204 Supplier Grainger Supplier No. 14M054
The Parker Reducing Bushing is made from 316 stainless steel and has NPT thread connections. It reduces pipe size from 1" to 3/4". The bushing has a pressure rating of 3000 psi at 100°F. It can be used with Schedule 80 pipe and is manufactured in the USA. The bushing measures 1.38" in length and weighs 0.19 lbs.