Learn More
Grainger Reducing Bushing 316 SS 3/4 x 1/2 in
Catalog No. 502752949
Encompass_Preferred
The Parker Reducing Bushing is made from 316 stainless steel and has NPT thread connections. It reduces pipe size from 1/2" to 1/4". The bushing has a pressure rating of 3000 psi at 100°F. It can be used with liquids, gases, and steam. The bushing dimensions are 1-1/2" length x 1" hex and it weighs 0.19 lbs. This stainless steel reducing bushing converts larger to smaller thread sizes while maintaining high pressure capabilities.
Catalog No. 50-275-2949 Supplier Grainger Supplier No. 14M056
The Parker Reducing Bushing is made from 316 stainless steel and has NPT thread connections. It reduces pipe size from 1/2" to 1/4". The bushing has a pressure rating of 3000 psi at 100°F. It can be used with liquids, gases, and steam. The bushing dimensions are 1-1/2" length x 1" hex and it weighs 0.19 lbs. This stainless steel reducing bushing converts larger to smaller thread sizes while maintaining high pressure capabilities.