Learn More
Grainger ELDON JAMES, Reducing Coupler, HDPE
Catalog No. NC2229013
Encompass_Preferred
The Eldon James Reducing Coupler HDPE is a plastic tubing fitting that allows you to connect tubing of two different sizes. It is made from HDPE plastic and has barbed ends to grip the tubing. The coupler has a 1/4" barb on one end that fits 1/4" ID tubing, and a 3/8" barb on the other end that fits 3/8" ID tubing. This allows you to make a transition between the two tubing sizes. The HDPE plastic construction makes it suitable for use in applications like air and water flow. Overall, this coupler provides a simple way to connect tubing of different diameters in industrial and commercial applications.
This item is not returnable. View return policy
Catalog No. NC2229013 Supplier Grainger Supplier No. 1ZJY2
The Eldon James Reducing Coupler HDPE is a plastic tubing fitting that allows you to connect tubing of two different sizes. It is made from HDPE plastic and has barbed ends to grip the tubing. The coupler has a 1/4" barb on one end that fits 1/4" ID tubing, and a 3/8" barb on the other end that fits 3/8" ID tubing. This allows you to make a transition between the two tubing sizes. The HDPE plastic construction makes it suitable for use in applications like air and water flow. Overall, this coupler provides a simple way to connect tubing of different diameters in industrial and commercial applications.