Learn More
Grainger Reducing Coupling 3/4 x 1/2 In PEX White
Catalog No. 502765257
Encompass_Preferred
The John Guest Reducing Coupler is a plastic push-to-connect fitting that allows you to join tubing of different sizes. It is made of acetal copolymer plastic and has O-ring seals. The coupler has 1/4" and 3/8" ports and is rated for pressures up to 232 psi at 73°F. It can be used with a variety of tubing materials including polyethylene, nylon, polyurethane, PVC, and more. The reducing coupler provides a simple push-in connection without requiring clamps, threads or tools. It is ideal for reducing tubing size in fluid transfer or pneumatic applications. The fitting is manufactured in the United Kingdom by John Guest, a leading producer of push-to-connect fittings.
Catalog No. 50-276-5257 Supplier Grainger Supplier No. 16T742
The John Guest Reducing Coupler is a plastic push-to-connect fitting that allows you to join tubing of different sizes. It is made of acetal copolymer plastic and has O-ring seals. The coupler has 1/4" and 3/8" ports and is rated for pressures up to 232 psi at 73°F. It can be used with a variety of tubing materials including polyethylene, nylon, polyurethane, PVC, and more. The reducing coupler provides a simple push-in connection without requiring clamps, threads or tools. It is ideal for reducing tubing size in fluid transfer or pneumatic applications. The fitting is manufactured in the United Kingdom by John Guest, a leading producer of push-to-connect fittings.