Learn More
Grainger Reducing Hex Coupling 316 SS 3/4 x 1/4
Supplier:  Grainger 14M051
Encompass_Preferred
The Parker Reducing Hex Coupling 316 is a 316 stainless steel hex coupling that reduces pipe size. It has a pressure rating of 3000 psi at 70°F and is made of 316 stainless steel for corrosion resistance. The coupling has female NPT threads on both ends and reduces pipe size from 1/2" to 3/8". It can be used to transition between different pipe sizes in hydraulic, pneumatic, lubrication, and industrial applications. The coupling body length is 1-1/4" and the hex dimensions are 7/8". It meets SAE J514 specifications. This reducing hex coupling provides a way to connect different pipe sizes in corrosive environments.
Catalog No. 50-275-2736