Learn More
Hach Company Reducing Solution, ORP Electrodes
Supplier:  Hach Company 2965349
Encompass
Used to condition ORP electrode surfaces for use in reducing sample matrices.
Catalog No. 50-199-6961