Learn More
Reducing Solution, Hydrazine - Copper Sulfate, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Hydrazine - Copper Sulfate |
| Color | Colorless to Light Blue Liquid |
| CAS | 7732-18-5, 10034-93-2, 7758-99-8 |
| CAS Max % | 99.9, 0.1, 0.002 |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| Quantity | 1 L |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.