Learn More
Grainger Reducing Tee 1/2 x1/2x3/4 In CTS PEX Wht
Catalog No. 502768438
Encompass_Preferred
The Grainger Approved Reducing Tee is a 3-piece fitting that connects pipes of different sizes. It is made of PVC plastic and has socket end connections. This tee allows you to transition from a larger to smaller pipe size while maintaining flow. It is ideal for DWV and pressure applications. The reducing tee has a maximum temperature rating of 140°F and maximum pressure rating of 480 PSI. It is certified by NSF International for use with potable water. This PVC reducing tee comes in sizes ranging from 1-1/2" x 1-1/4" to 4" x 3" and is available in both sch 40 and sch 80. The fittings are designed for use with PVC, CPVC, and ABS pipe and fittings.
Catalog No. 50-276-8438 Supplier Grainger Supplier No. 16T749
The Grainger Approved Reducing Tee is a 3-piece fitting that connects pipes of different sizes. It is made of PVC plastic and has socket end connections. This tee allows you to transition from a larger to smaller pipe size while maintaining flow. It is ideal for DWV and pressure applications. The reducing tee has a maximum temperature rating of 140°F and maximum pressure rating of 480 PSI. It is certified by NSF International for use with potable water. This PVC reducing tee comes in sizes ranging from 1-1/2" x 1-1/4" to 4" x 3" and is available in both sch 40 and sch 80. The fittings are designed for use with PVC, CPVC, and ABS pipe and fittings.