Learn More
Grainger Reducing Tee 3/4 x1/2x3/4 In CTS PEX Wht
Catalog No. 502767646
Encompass_Preferred
The John Guest Reducing Tee Plastic is a reducing tee fitting made from polypropylene for use with plastic tubing. It has 1/2" and 3/8" ports and is suitable for pressures up to 100 psi at temperatures up to 180°F. The tee has a maximum flow rate of 4.5 gpm and allows a change in tubing size while maintaining flow. It can be used for connecting plastic tubing in plumbing, irrigation, water filtration, beverage dispensing, and other fluid transfer applications. The tee is made in the USA and is lead-free, corrosion resistant, and durable. It provides a reliable connection method for reducing tubing size in plastic tubing systems.
Catalog No. 50-276-7646 Supplier Grainger Supplier No. 16T750
The John Guest Reducing Tee Plastic is a reducing tee fitting made from polypropylene for use with plastic tubing. It has 1/2" and 3/8" ports and is suitable for pressures up to 100 psi at temperatures up to 180°F. The tee has a maximum flow rate of 4.5 gpm and allows a change in tubing size while maintaining flow. It can be used for connecting plastic tubing in plumbing, irrigation, water filtration, beverage dispensing, and other fluid transfer applications. The tee is made in the USA and is lead-free, corrosion resistant, and durable. It provides a reliable connection method for reducing tubing size in plastic tubing systems.