Learn More
Grainger Reducing Tee 3/4x3/4 x1/2 In CTS PEX Wht
Catalog No. 502767649
Encompass_Preferred
The John Guest Reducing Tee Plastic is a reducing tee fitting made from polypropylene for use with plastic tubing. It has 1/2" and 3/8" ports and is suitable for pressures up to 232 PSI at 73°F. The tee has a maximum temperature rating of 180°F and is NSF/ANSI 61 certified for potable water applications. This product allows a transition from larger to smaller tubing sizes. It can be used for water lines, beverage dispensing, air lines, chemical transfer, and other fluid transfer applications. The tee fitting is made in the USA.
Catalog No. 50-276-7649 Supplier Grainger Supplier No. 16T747
The John Guest Reducing Tee Plastic is a reducing tee fitting made from polypropylene for use with plastic tubing. It has 1/2" and 3/8" ports and is suitable for pressures up to 232 PSI at 73°F. The tee has a maximum temperature rating of 180°F and is NSF/ANSI 61 certified for potable water applications. This product allows a transition from larger to smaller tubing sizes. It can be used for water lines, beverage dispensing, air lines, chemical transfer, and other fluid transfer applications. The tee fitting is made in the USA.