Learn More
DSI LLC Reducing union 1/16" to 360um, 150um bore, for PEEK+FS tubing (for<10kpsi)
Catalog No. 502255047
Encompass_Preferred
Reducing union 1/16" to 360um, 150um bore, for PEEK+FS tubing (for<10kpsi)
Catalog No. 50-225-5047 Supplier DSI LLC Supplier No. JRC360RU1PK6
Reducing union 1/16" to 360um, 150um bore, for PEEK+FS tubing (for<10kpsi)