Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Reduction Coupler with 5/8 in. ID x 5/16 in. ID - Natural Kynar - QC
SDP

Catalog No. 01000573 Shop All Fisherbrand Products
This item is not returnable. View return policy
Catalog No. 01-000-573
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing, for a tube-to-tube connection.

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661
  • USP81
  • USP 8
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
Material Natural Kynar
For Use With (Application) Tube to Tube Connector for Medical, Pharmaceutical, Industrial applications
Packaging Quantity 100 Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Reduction Coupler
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.