Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC REDV TFA | >99.81% | 517.53 | 25 MG
SDP

Catalog No. 5000397411
Encompass_Preferred

REDV TFA is the minimal active sequence within the CS5 site of the alternatively spliced type III connecting segment (IIICS) region of fibronectin. It mediates adhesion to the IIICS region of plasma fibronectin by binding the integrin alpha 4 beta 1 (α4β1). This peptide can be utilized in the research of cell adhesion.

  • Target: Integrin
  • Pathway: Cytoskeleton
  • Storage (powder): -80°C for 2 years, -20°C for 1 year (sealed, away from moisture and light)
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light)
  • Solubility (in vitro): H₂O: 100 mg/mL (requires ultrasonic)
  • Biological activity: Mediates adhesion to the IIICS region of plasma fibronectin by binding the integrin alpha 4 beta 1(a4β1)

Catalog No. 50-003-97411 Supplier Medchemexpress LLC Supplier No. 25MGHYP3522A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

REDV TFA is the minimal active sequence within the CS5 site of the alternatively spliced type III connecting segment (IIICS) region of fibronectin. It mediates adhesion to the IIICS region of plasma fibronectin by binding the integrin alpha 4 beta 1 (α4β1). This peptide can be utilized in the research of cell adhesion.

  • Target: Integrin
  • Pathway: Cytoskeleton
  • Storage (powder): -80°C for 2 years, -20°C for 1 year (sealed, away from moisture and light)
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light)
  • Solubility (in vitro): H₂O: 100 mg/mL (requires ultrasonic)
  • Biological activity: Mediates adhesion to the IIICS region of plasma fibronectin by binding the integrin alpha 4 beta 1(a4β1)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.