Learn More
MSC REED Instruments Combustible Gas Leak Detector, 5 ppm, 1 ea
Catalog No. 5000769760
Encompass
This combustible gas leak detector identifies a wide range of combustible gases with high accuracy, detecting leaks as small as 5 ppm of gasoline. It features a 15 in gooseneck for accessing confined spaces and provides both audible and visual alarm indicators, making it suitable for residential, commercial, and industrial safety applications.
  • Detects a wide range of combustible gases
  • Quick response and high accuracy detects small leaks
  • Features a 15 in (390 mm) gooseneck for hard-to-reach areas
  • User adjustable sensitivity (High/Low)
  • Audible (buzzer) and visual (LED) alarm indicators
  • User replaceable sensor
  • Includes reference leak source, batteries, and hard carrying case
Catalog No. 50-007-69760 Supplier MSC Supplier No. 10164127
This combustible gas leak detector identifies a wide range of combustible gases with high accuracy, detecting leaks as small as 5 ppm of gasoline. It features a 15 in gooseneck for accessing confined spaces and provides both audible and visual alarm indicators, making it suitable for residential, commercial, and industrial safety applications.
  • Detects a wide range of combustible gases
  • Quick response and high accuracy detects small leaks
  • Features a 15 in (390 mm) gooseneck for hard-to-reach areas
  • User adjustable sensitivity (High/Low)
  • Audible (buzzer) and visual (LED) alarm indicators
  • User replaceable sensor
  • Includes reference leak source, batteries, and hard carrying case