Learn More
MSC REED Instruments Coupling Gel, Ultrasonic, Water Soluble, Slow Drying, 8.45 oz
Catalog No. 5000513118
Encompass
This coupling gel is designed for use with ultrasonic thickness gages. It is slow drying and water soluble, providing good transducer lubrication for accurate measurements.
  • Slow drying with good transducer lubrication
  • Water soluble
  • Includes applicator bottle
  • 8.45 fl oz (250 ml) capacity
Catalog No. 50-005-13118 Supplier MSC Supplier No. 10166908
This coupling gel is designed for use with ultrasonic thickness gages. It is slow drying and water soluble, providing good transducer lubrication for accurate measurements.
  • Slow drying with good transducer lubrication
  • Water soluble
  • Includes applicator bottle
  • 8.45 fl oz (250 ml) capacity